Assay Details
Target Gene Details
Entrez Gene ID: | 4881 |
Gene Name: | natriuretic peptide receptor 1 |
Gene Aliases: |
ANPRA, ANPa, GUC2A, GUCY2A, NPRA |
Location: |
Chr.1:153678617-153693992 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPR1 | NM_000906.3 | NP_000897.3 | ||
XM_005245218.1 | XP_005245275.1 | |||
AK025024.1 | ||||
AK298090.1 | ||||
AK300446.1 | ||||
BC063304.1 | AAH63304.1 | |||
S72628.1 | AAD14112.1 | |||
X15357.1 | CAA33417.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv464028 | Chr.1:153649744 - 153750561 on Build GRCh38 | Gain | INTS3 ILF2 SNAPIN MIR8083 NPR1 |
dgv582n54 | Chr.1:153639657 - 153750561 on Build GRCh38 | Gain | INTS3 ILF2 CHTOP SNAPIN MIR8083 NPR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |