Assay Details
Target Gene Details
Entrez Gene ID: | 2948 |
Gene Name: | glutathione S-transferase mu 4 |
Gene Aliases: |
GSTM4-4, GTM4 |
Location: |
Chr.1:109656076-109665501 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSTM4 | NM_147148.2 | NP_671489.1 | ||
AI949679.1 | ||||
BI791533.1 | ||||
M99422.1 | AAA58623.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv258n100 | Chr.1:109658958 - 109700319 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv257n100 | Chr.1:109650144 - 109707602 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv254n100 | Chr.1:109631914 - 109700319 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
dgv15n111 | Chr.1:109664699 - 109703504 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
dgv77e212 | Chr.1:109635559 - 109709330 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv259n100 | Chr.1:109663341 - 109700319 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |
nsv547486 | Chr.1:109657738 - 109690459 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv256n100 | Chr.1:109638683 - 109700319 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv25172 | Chr.1:109644224 - 109716486 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
esv3587153 | Chr.1:109660894 - 109674476 on Build GRCh38 | Loss |
![]() ![]() |
dgv253n100 | Chr.1:109613590 - 109700319 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)