Assay Details
Target Gene Details
Entrez Gene ID: | 101928627 |
Gene Name: | uncharacterized LOC101928627 |
Gene Aliases: |
- |
Location: |
Chr.X:36365626-36440292 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928627 | NR_110412.1 | |||
AK092146.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470349 | Chr.X:35988125 - 36420457 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)