Assay Details
Target Gene Details
Entrez Gene ID: | 140462 |
Gene Name: | ankyrin repeat and SOCS box containing 9 |
Gene Aliases: |
- |
Location: |
Chr.X:15243987-15270476 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASB9 | NM_001031739.2 | NP_001026909.1 | ||
NM_001168530.1 | NP_001162002.1 | |||
NM_001168531.1 | NP_001162003.1 | |||
NM_024087.2 | NP_076992.1 | |||
XM_005274446.1 | XP_005274503.1 | |||
XM_011545458.1 | XP_011543760.1 | |||
XM_011545460.1 | XP_011543762.1 | |||
XM_017029283.1 | XP_016884772.1 | |||
XM_017029284.1 | XP_016884773.1 | |||
XM_017029285.1 | XP_016884774.1 | |||
XM_017029286.1 | XP_016884775.1 | |||
AK000643.1 | BAA91302.1 | |||
AK292270.1 | ||||
AL080091.2 | CAB45706.2 | |||
AY337578.1 | AAQ17469.1 | |||
BC001244.1 | AAH01244.1 | |||
BC013172.1 | AAH13172.1 | |||
DA943920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528005 | Chr.X:15247097 - 15264373 on Build GRCh38 | Gain | ASB9 |
dgv2226e212 | Chr.X:15248328 - 15272357 on Build GRCh38 | Loss | ASB9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |