Assay Details
Target Gene Details
Entrez Gene ID: | 1536 |
Gene Name: | cytochrome b-245 beta chain |
Gene Aliases: |
AMCBX2, CGD, GP91-1, GP91-PHOX, GP91PHOX, IMD34, NOX2, p91-PHOX |
Location: |
Chr.X:37780017-37813461 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYBB | NM_000397.3 | NP_000388.2 | ||
AK289753.1 | ||||
AK301476.1 | ||||
AK301491.1 | ||||
AK304033.1 | ||||
AK312929.1 | ||||
BC032720.1 | AAH32720.1 | |||
DA995835.1 | ||||
KU178009.1 | ||||
KU178010.1 | ||||
X04011.1 | CAA27635.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |