Assay Details
Target Gene Details
Entrez Gene ID: | 79135 |
Gene Name: | apolipoprotein O |
Gene Aliases: |
FAM121B, MIC26, Mic23, My025 |
Location: |
Chr.X:23833348-23907940 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOO | NM_024122.4 | NP_077027.1 | ||
NR_026545.2 | ||||
XM_011545584.2 | XP_011543886.1 | |||
XM_011545585.2 | XP_011543887.1 | |||
XM_011545586.2 | XP_011543888.1 | |||
XM_011545587.2 | XP_011543889.1 | |||
XM_017029837.1 | XP_016885326.1 | |||
AF061264.1 | AAG43139.1 | |||
AK311865.1 | ||||
AY359114.1 | AAQ89472.1 | |||
BC002333.2 | AAH02333.1 | |||
BC010102.2 | AAH10102.1 | |||
BC016814.1 | AAH16814.1 | |||
BX099786.1 | ||||
EL951748.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |