Assay Details
Target Gene Details
Entrez Gene ID: | 55634 |
Gene Name: | KRAB box domain containing 4 |
Gene Aliases: |
ZNF673 |
Location: |
Chr.X:46447189-46474639 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KRBOX4 | NM_001129898.1 | NP_001123370.1 | ||
NM_001129899.1 | NP_001123371.1 | |||
NM_001129900.1 | NP_001123372.1 | |||
NM_017776.2 | NP_060246.2 | |||
AK000351.1 | BAA91104.1 | |||
AK096413.1 | ||||
AK289703.1 | ||||
BC012569.1 | AAH12569.1 | |||
DA152034.1 | ||||
DA631861.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576854 | Chr.X:46457004 - 46479835 on Build GRCh38 | Gain | KRBOX4 |
nsv526411 | Chr.X:45810991 - 46476384 on Build GRCh38 | Gain | KRBOX4 LOC401585 LINC01186 |
esv3576853 | Chr.X:46169680 - 46644654 on Build GRCh38 | Gain | ZNF674 CHST7 KRBOX4 SLC9A7 LINC01186 ZNF674-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |