Assay Details
Target Gene Details
Entrez Gene ID: | 5251 |
Gene Name: | phosphate regulating endopeptidase homolog, X-linked |
Gene Aliases: |
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH |
Location: |
Chr.X:22032327-22251310 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHEX | NM_000444.5 | NP_000435.3 | ||
NM_001282754.1 | NP_001269683.1 | |||
XM_011545533.1 | XP_011543835.1 | |||
XM_011545536.2 | XP_011543838.1 | |||
XM_017029579.1 | XP_016885068.1 | |||
AD000712.1 | AAB51604.1 | |||
AK298030.1 | ||||
AK301528.1 | ||||
AK304551.1 | ||||
BC105057.1 | AAI05058.1 | |||
BC105059.1 | AAI05060.1 | |||
U60475.1 | AAC50552.1 | |||
U75645.1 | AAB47749.1 | |||
U82970.1 | AAC24487.1 | |||
U87284.1 | AAB47562.1 |
Target Gene Details
Entrez Gene ID: | 100873065 |
Gene Name: | PTCHD1 antisense RNA (head to head) |
Gene Aliases: |
DDX53-AS1, PTCHD1AS1, PTCHD1AS2 |
Location: |
Chr.X:22193005-23293146 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTCHD1-AS | NR_073010.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521384 | Chr.X:22198650 - 22953327 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)