Assay Details
Target Gene Details
Entrez Gene ID: | 100303728 |
Gene Name: | SLC25A5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:119466033-119469120 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A5-AS1 | NR_028443.1 | 1 | 1008 | |
NR_134914.1 | ||||
NR_134915.1 | ||||
AK092094.1 | 1 | 984 | BAC03803.1 | |
AK094280.1 |