Assay Details
Target Gene Details
Entrez Gene ID: | 169966 |
Gene Name: | family with sequence similarity 46 member D |
Gene Aliases: |
CT1.26, CT112 |
Location: |
Chr.X:80335504-80445311 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM46D | NM_001170574.1 | NP_001164045.1 | ||
NM_152630.4 | NP_689843.1 | |||
AK128147.1 | ||||
AK313875.1 | ||||
BC028710.2 | AAH28710.1 | |||
BC030784.2 | AAH30784.1 | |||
BC034979.2 | AAH34979.1 | |||
BX506299.1 | ||||
BX537938.1 | CAD97909.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)