Assay Details
Target Gene Details
Entrez Gene ID: | 340562 |
Gene Name: | spermidine/spermine N1-acetyl transferase-like 1 |
Gene Aliases: |
- |
Location: |
Chr.X:85092286-85116229 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SATL1 | XM_011530940.2 | XP_011529242.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |