Assay Details
Target Gene Details
Entrez Gene ID: | 7102 |
Gene Name: | tetraspanin 7 |
Gene Aliases: |
A15, CCG-B7, CD231, DXS1692E, MRX58, MXS1, TALLA-1, TM4SF2, TM4SF2b |
Location: |
Chr.X:38561478-38688918 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSPAN7 | NM_004615.3 | NP_004606.2 | ||
AB062057.1 | BAB55824.1 | |||
AB102665.1 | BAC81134.1 | |||
AK293178.1 | ||||
AK293534.1 | ||||
AK293552.1 | ||||
AK312343.1 | ||||
BC018036.1 | AAH18036.1 | |||
D10653.1 | BAA01501.1 | |||
D29808.1 | BAA06191.1 | |||
DB475293.1 | ||||
L10373.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs137925146] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)