Assay Details
Target Gene Details
Entrez Gene ID: | 257397 |
Gene Name: | TGF-beta activated kinase 1/MAP3K7 binding protein 3 |
Gene Aliases: |
MAP3K7IP3, NAP1 |
Location: |
Chr.X:30827442-30889394 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TAB3 | NM_152787.4 | NP_690000.3 | ||
XM_005274483.3 | XP_005274540.2 | |||
XM_005274484.2 | XP_005274541.2 | |||
XM_017029399.1 | 10 | 2910 | XP_016884888.1 | |
XM_017029400.1 | 10 | 2765 | XP_016884889.1 | |
XM_017029401.1 | XP_016884890.1 | |||
AK297274.1 | ||||
AL832071.1 | 9 | 6150 | CAI46137.1 | |
AY331591.1 | AAQ92938.1 | |||
AY331592.1 | AAQ92939.1 | |||
AY371491.1 | AAQ88279.1 | |||
AY437560.1 | AAR06179.1 | |||
BC032526.1 | AAH32526.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |