Assay Details
Target Gene Details
Entrez Gene ID: | 139285 |
Gene Name: | APC membrane recruitment protein 1 |
Gene Aliases: |
FAM123B, OSCS, WTX |
Location: |
Chr.X:64185117-64205744 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMER1 | NM_152424.3 | NP_689637.3 | ||
AK097146.1 | BAC04964.1 | |||
DA939151.1 | ||||
EF186024.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521560 | Chr.X:64063724 - 64245972 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)