Assay Details
Target Gene Details
Entrez Gene ID: | 5251 |
Gene Name: | phosphate regulating endopeptidase homolog, X-linked |
Gene Aliases: |
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH |
Location: |
Chr.X:22032327-22251310 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHEX | NM_000444.5 | NP_000435.3 | ||
NM_001282754.1 | NP_001269683.1 | |||
AD000712.1 | AAB51604.1 | |||
AK298030.1 | ||||
BC105057.1 | AAI05058.1 | |||
BC105059.1 | AAI05060.1 | |||
U60475.1 | AAC50552.1 | |||
U75645.1 | AAB47749.1 | |||
U82970.1 | AAC24487.1 | |||
U87284.1 | AAB47562.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |