Assay Details
Target Gene Details
Entrez Gene ID: | 25975 |
Gene Name: | EGF like domain multiple 6 |
Gene Aliases: |
MAEG, W80 |
Location: |
Chr.X:13569575-13633575 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EGFL6 | NM_001167890.1 | NP_001161362.1 | ||
NM_015507.3 | NP_056322.2 | |||
AF186084.1 | AAF27812.1 | |||
AF193055.1 | AAG22483.1 | |||
AJ245671.1 | CAB92132.1 | |||
AK075214.1 | BAC11477.1 | |||
AK300137.1 | ||||
AK300236.1 | ||||
AK315018.1 | ||||
AL117610.1 | CAB56014.2 | |||
AY358333.1 | AAQ88699.1 | |||
BC038587.1 | AAH38587.1 | |||
DA534733.1 | ||||
DC380958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576773 | Chr.X:13071156 - 13600068 on Build GRCh38 | Gain | LOC105373133 LINC01203 GPX1P1 GS1-600G8.3 ATXN3L MIR6086 EGFL6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |