Assay Details
Target Gene Details
Entrez Gene ID: | 5880 |
Gene Name: | ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2) |
Gene Aliases: |
EN-7, Gx, HSPC022, p21-Rac2 |
Location: |
Chr.22:37225261-37244299 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAC2 | NM_002872.4 | NP_002863.1 | ||
XM_006724286.3 | XP_006724349.1 | |||
AF498965.1 | AAM21112.1 | |||
BC001485.2 | AAH01485.1 | |||
BI908751.1 | ||||
BQ934070.1 | ||||
BT006919.1 | AAP35565.1 | |||
CR456555.1 | CAG30441.1 | |||
M29871.1 | AAA36538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834188 | Chr.22:37210658 - 37396179 on Build GRCh38 | Loss | ELFN2 RAC2 LOC100506271 CYTH4 SSTR3 |
nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss | TST LGALS2 TRIOBP ELFN2 RAC2 LOC100506271 GGA1 MPST MFNG LOC102724378 CARD10 SSTR3 NOL12 IL2RB TMPRSS6 CDC42EP1 C1QTNF6 CYTH4 KCTD17 TEX33 LGALS1 LOC101927051 PDXP SH3BP1 |
nsv459883 | Chr.22:37215767 - 37242920 on Build GRCh38 | Loss | RAC2 SSTR3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |