Assay Details
Target Gene Details
Entrez Gene ID: | 23786 |
Gene Name: | BCL2 like 13 |
Gene Aliases: |
BCL-RAMBO, Bcl2-L-13, MIL1 |
Location: |
Chr.22:17628855-17730855 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss | LOC101929372 CECR2 BCL2L13 SLC25A18 ATP6V1E1 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | BID LINC00528 LOC101929372 CECR2 MIR3198-1 MICAL3 BCL2L13 SLC25A18 ATP6V1E1 |
esv3893429 | Chr.22:17600825 - 17678777 on Build GRCh38 | Gain | BCL2L13 ATP6V1E1 |
esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss | BID LINC00528 LOC101929372 FLJ41941 MIR648 MIR3198-1 MICAL3 BCL2L13 SLC25A18 ATP6V1E1 |
dgv4465n100 | Chr.22:17577691 - 17706530 on Build GRCh38 | Gain | LOC101929372 BCL2L13 SLC25A18 ATP6V1E1 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | MIR3198-1 MICAL3 LOC102725072 LOC100996415 TUBA8 BID LINC00528 FLJ41941 MIR648 TMEM191B RIMBP3 USP18 LOC100996401 BCL2L13 SLC25A18 PI4KAP1 ATP6V1E1 LOC642643 LOC101929372 GGT3P CECR2 LOC100996432 LOC729461 PEX26 FAM230A |
nsv834119 | Chr.22:17665133 - 17795994 on Build GRCh38 | Loss | BID LINC00528 MIR3198-1 MICAL3 BCL2L13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |