Assay Details
Target Gene Details
Entrez Gene ID: | 80831 |
Gene Name: | apolipoprotein L5 |
Gene Aliases: |
APOL-V, APOLV |
Location: |
Chr.22:35708424-35730279 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOL5 | NM_030642.1 | NP_085145.1 | ||
XM_006724321.3 | XP_006724384.1 | |||
XM_017028945.1 | XP_016884434.1 | |||
AY014878.1 | AAK07723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3612 | Chr.22:35713114 - 35761709 on Build GRCh38 | Insertion | APOL5 RBFOX2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |