Assay Details
Target Gene Details
Entrez Gene ID: | 10478 |
Gene Name: | solute carrier family 25 member 17 |
Gene Aliases: |
PMP34 |
Location: |
Chr.22:40769630-40819399 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A17 | NM_001282726.1 | NP_001269655.1 | ||
NM_001282727.1 | NP_001269656.1 | |||
NM_006358.3 | NP_006349.1 | |||
NR_104235.1 | ||||
NR_104236.1 | ||||
NR_104237.1 | ||||
NR_104238.1 | ||||
AK292924.1 | ||||
AK298215.1 | ||||
AK300553.1 | ||||
AU123445.1 | ||||
BC005957.1 | AAH05957.1 | |||
BC012998.2 | AAH12998.1 | |||
BQ440957.1 | ||||
BU595416.1 | ||||
BX352732.2 | ||||
BX647991.1 | ||||
CR456577.1 | CAG30463.1 | |||
DC397321.1 | ||||
Y12860.1 | CAA73367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834206 | Chr.22:40696722 - 40838881 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1062438 | Chr.22:40646389 - 40806134 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)