Assay Details
Target Gene Details
Entrez Gene ID: | 3162 |
Gene Name: | heme oxygenase 1 |
Gene Aliases: |
HMOX1D, HO-1, HSP32, bK286B10 |
Location: |
Chr.22:35381067-35394214 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HMOX1 | NM_002133.2 | NP_002124.1 | ||
AK313120.1 | ||||
BC001491.2 | AAH01491.1 | |||
BP288767.1 | ||||
CR456505.1 | CAG30391.1 | |||
CR541945.1 | CAG46743.1 | |||
CR541968.1 | CAG46766.1 | |||
GU187011.1 | ||||
GU187012.1 | ||||
JF323038.1 | ||||
M23041.1 | AAA50403.1 | |||
X06985.1 | CAA30045.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3611 | Chr.22:35363383 - 35385153 on Build GRCh38 | Insertion | HMOX1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |