Assay Details
Target Gene Details
Entrez Gene ID: | 100874197 |
Gene Name: | EFCAB6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:43516254-43536605 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EFCAB6-AS1 | NR_046563.1 | |||
AK090843.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv589098 | Chr.22:43463583 - 43718462 on Build GRCh38 | Gain | EFCAB6 MPPED1 EFCAB6-AS1 |
nsv471205 | Chr.22:43467417 - 43718981 on Build GRCh38 | Gain | EFCAB6 MPPED1 EFCAB6-AS1 |