Assay Details
Target Gene Details
Entrez Gene ID: | 23786 |
Gene Name: | BCL2 like 13 |
Gene Aliases: |
BCL-RAMBO, Bcl2-L-13, MIL1 |
Location: |
Chr.22:17628855-17730855 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2669345 | Chr.22:17643887 - 17646733 on Build GRCh38 | Deletion | BCL2L13 |
nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss | CECR2 LOC101929372 SLC25A18 ATP6V1E1 BCL2L13 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | CECR2 BID LOC101929372 MICAL3 SLC25A18 ATP6V1E1 LINC00528 BCL2L13 MIR3198-1 |
dgv2429n106 | Chr.22:17643774 - 17647145 on Build GRCh38 | Deletion | BCL2L13 |
esv3893429 | Chr.22:17600825 - 17678777 on Build GRCh38 | Gain | ATP6V1E1 BCL2L13 |
esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss | BID LOC101929372 MICAL3 SLC25A18 FLJ41941 ATP6V1E1 LINC00528 BCL2L13 MIR3198-1 MIR648 |
dgv4465n100 | Chr.22:17577691 - 17706530 on Build GRCh38 | Gain | LOC101929372 SLC25A18 ATP6V1E1 BCL2L13 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | LOC729461 ATP6V1E1 BCL2L13 MIR648 SLC25A18 GGT3P MIR3198-1 PEX26 PI4KAP1 TMEM191B CECR2 LOC100996415 RIMBP3 LOC100996432 LOC642643 USP18 LOC100996401 LINC00528 TUBA8 BID LOC101929372 MICAL3 LOC102725072 FLJ41941 FAM230A |
esv3647229 | Chr.22:17643640 - 17646896 on Build GRCh38 | Loss | BCL2L13 |
esv22768 | Chr.22:17644216 - 17646684 on Build GRCh38 | Loss | BCL2L13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |