Assay Details
Target Gene Details
Entrez Gene ID: | 5594 |
Gene Name: | mitogen-activated protein kinase 1 |
Gene Aliases: |
ERK, ERK-2, ERK2, ERT1, MAPK2, P42MAPK, PRKM1, PRKM2, p38, p40, p41, p41mapk, p42-MAPK |
Location: |
Chr.22:21759657-21867680 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAPK1 | NM_002745.4 | NP_002736.3 | ||
AK090869.1 | ||||
AK295188.1 | ||||
AK307615.1 | ||||
BC099905.1 | AAH99905.1 | |||
EF565108.1 | ||||
Z11694.1 | CAA77752.1 | |||
Z11695.1 | CAA77753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv588322 | Chr.22:21732084 - 21832190 on Build GRCh38 | Loss | MAPK1 YPEL1 |
nsv510804 | Chr.22:21670242 - 21778475 on Build GRCh38 | Deletion | PPIL2 MAPK1 YPEL1 |
nsv1060548 | Chr.22:21724874 - 21772537 on Build GRCh38 | Gain | MAPK1 YPEL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |