Assay Details
Target Gene Details
Entrez Gene ID: | 157 |
Gene Name: | G protein-coupled receptor kinase 3 |
Gene Aliases: |
ADRBK2, BARK2 |
Location: |
Chr.22:25564894-25729294 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRK3 | NM_005160.3 | NP_005151.2 | ||
AK058103.1 | ||||
AK292234.1 | ||||
AK310398.1 | ||||
BC036797.1 | AAH36797.1 | |||
BX387339.2 | ||||
X69117.1 | CAA48870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066595 | Chr.22:25596614 - 26071416 on Build GRCh38 | Gain |
![]() ![]() |
nsv517205 | Chr.22:25244661 - 25629591 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1058343 | Chr.22:25507736 - 25642811 on Build GRCh38 | Loss |
![]() |
esv2758829 | Chr.22:24970834 - 25650546 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2751952 | Chr.22:25262579 - 25754679 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3647478 | Chr.22:25598397 - 25702971 on Build GRCh38 | Gain |
![]() |
nsv469691 | Chr.22:25457547 - 25622123 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |
nsv471598 | Chr.22:25457548 - 25622123 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)