Assay Details
Target Gene Details
Entrez Gene ID: | 23786 |
Gene Name: | BCL2 like 13 |
Gene Aliases: |
BCL-RAMBO, Bcl2-L-13, MIL1 |
Location: |
Chr.22:17628855-17730855 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2665896 | Chr.22:17642077 - 17643673 on Build GRCh38 | Deletion | BCL2L13 |
nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss | LOC101929372 ATP6V1E1 SLC25A18 CECR2 BCL2L13 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | LOC101929372 BID ATP6V1E1 MICAL3 LINC00528 MIR3198-1 SLC25A18 CECR2 BCL2L13 |
esv3893429 | Chr.22:17600825 - 17678777 on Build GRCh38 | Gain | ATP6V1E1 BCL2L13 |
esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss | LOC101929372 BID MIR648 ATP6V1E1 FLJ41941 MICAL3 LINC00528 MIR3198-1 SLC25A18 BCL2L13 |
dgv4465n100 | Chr.22:17577691 - 17706530 on Build GRCh38 | Gain | LOC101929372 ATP6V1E1 SLC25A18 BCL2L13 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | BID LOC100996401 MICAL3 USP18 BCL2L13 FAM230A GGT3P LOC729461 TUBA8 TMEM191B LOC102725072 LINC00528 CECR2 LOC101929372 MIR648 LOC642643 ATP6V1E1 LOC100996415 FLJ41941 RIMBP3 MIR3198-1 PI4KAP1 SLC25A18 LOC100996432 PEX26 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |