Assay Details
Target Gene Details
Entrez Gene ID: | 10454 |
Gene Name: | TGF-beta activated kinase 1/MAP3K7 binding protein 1 |
Gene Aliases: |
3'-Tab1, MAP3K7IP1 |
Location: |
Chr.22:39399754-39437127 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TAB1 | NM_006116.2 | NP_006107.1 | ||
NM_153497.2 | NP_705717.1 | |||
AF425640.1 | AAN32760.1 | |||
AK291668.1 | ||||
AK296314.1 | ||||
AK308903.1 | ||||
AK312456.1 | ||||
AK316004.1 | ||||
BC050554.1 | AAH50554.1 | |||
U49928.1 | AAC12660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834199 | Chr.22:39324389 - 39531343 on Build GRCh38 | Loss | TAB1 MGAT3-AS1 RPS19BP1 MGAT3 LOC100506472 MIEF1 ATF4 SYNGR1 |
nsv834198 | Chr.22:39222748 - 39409818 on Build GRCh38 | Loss | SNORD139 SNORD83A SNORD83B TAB1 PDGFB RPL3 SNORD43 SYNGR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |