Assay Details
Target Gene Details
Entrez Gene ID: | 27351 |
Gene Name: | desumoylating isopeptidase 1 |
Gene Aliases: |
D15Wsu75e, DESI2, DJ347H13.4, DeSI-1, FAM152B, PPPDE2 |
Location: |
Chr.22:41598028-41621077 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DESI1 | NM_015704.2 | NP_056519.1 | ||
XM_005261571.4 | XP_005261628.1 | |||
AF038183.1 | ||||
AK090725.1 | ||||
BC093956.1 | AAH93956.1 | |||
BC112179.1 | AAI12180.1 | |||
BX403547.2 | ||||
CR456458.1 | CAG30344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055166 | Chr.22:41536399 - 41622240 on Build GRCh38 | Gain | LOC105373044 CSDC2 PMM1 XRCC6 POLR3H DESI1 |
nsv834209 | Chr.22:41563516 - 41746056 on Build GRCh38 | Loss | SNU13 MEI1 CSDC2 PMM1 XRCC6 C22orf46 DESI1 |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss | RANGAP1 ZC3H7B TEF EP300-AS1 SNORD140 LOC105373044 TOB2 LOC100996598 PMM1 C22orf46 MIR6889 PHF5A SNU13 EP300 ACO2 L3MBTL2 MEI1 CSDC2 XRCC6 POLR3H CHADL MIR1281 DESI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |