Assay Details
Target Gene Details
Entrez Gene ID: | 83733 |
Gene Name: | solute carrier family 25 member 18 |
Gene Aliases: |
GC2 |
Location: |
Chr.22:17562470-17590890 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3568230 | Chr.22:17570832 - 17577690 on Build GRCh38 | Loss | SLC25A18 |
nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss | CECR2 ATP6V1E1 SLC25A18 LOC101929372 BCL2L13 |
dgv4464n100 | Chr.22:17570790 - 17611340 on Build GRCh38 | Gain | ATP6V1E1 SLC25A18 LOC101929372 |
nsv1065055 | Chr.22:17559085 - 17632626 on Build GRCh38 | Gain | ATP6V1E1 SLC25A18 LOC101929372 BCL2L13 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | CECR2 ATP6V1E1 SLC25A18 MIR3198-1 LINC00528 BID LOC101929372 BCL2L13 MICAL3 |
nsv3553 | Chr.22:17553601 - 17608096 on Build GRCh38 | Deletion | CECR2 ATP6V1E1 SLC25A18 LOC101929372 |
nsv510802 | Chr.22:17550100 - 17604891 on Build GRCh38 | Deletion | CECR2 ATP6V1E1 SLC25A18 LOC101929372 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | LOC100996415 LOC101929372 MICAL3 FAM230A GGT3P LINC00528 PEX26 TUBA8 LOC100996401 USP18 LOC102725072 LOC100996432 SLC25A18 MIR648 LOC642643 BID BCL2L13 FLJ41941 TMEM191B CECR2 ATP6V1E1 MIR3198-1 PI4KAP1 RIMBP3 LOC729461 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |