Assay Details
Target Gene Details
Entrez Gene ID: | 2547 |
Gene Name: | X-ray repair cross complementing 6 |
Gene Aliases: |
CTC75, CTCBF, G22P1, KU70, ML8, TLAA |
Location: |
Chr.22:41621163-41664048 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC6 | NM_001288976.1 | NP_001275905.1 | ||
NM_001288977.1 | NP_001275906.1 | |||
NM_001288978.1 | NP_001275907.1 | |||
NM_001469.4 | NP_001460.1 | |||
AK055786.1 | ||||
AK293439.1 | ||||
AK304580.1 | ||||
AK315618.1 | ||||
BC008343.1 | AAH08343.1 | |||
BC010034.2 | AAH10034.1 | |||
BC012154.2 | AAH12154.1 | |||
BC018259.2 | AAH18259.1 | |||
BC072449.1 | AAH72449.1 | |||
BQ217843.1 | ||||
BU902323.1 | ||||
CR456492.1 | CAG30378.1 | |||
CR542219.1 | CAG47015.1 | |||
J04607.1 | AAA61177.1 | |||
J04611.1 | AAA51733.1 | |||
M32865.1 | AAA36155.1 | |||
S38729.1 | AAB22381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834209 | Chr.22:41563516 - 41746056 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)