Assay Details
Target Gene Details
Entrez Gene ID: | 23541 |
Gene Name: | SEC14 like lipid binding 2 |
Gene Aliases: |
C22orf6, SPF, TAP, TAP1 |
Location: |
Chr.22:30396941-30425303 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEC14L2 | NM_001204204.2 | NP_001191133.1 | ||
NM_001291932.1 | NP_001278861.1 | |||
NM_012429.4 | NP_036561.1 | |||
NM_033382.2 | NP_203740.1 | |||
AB033012.2 | ||||
AK091402.1 | ||||
AK223587.1 | BAD97307.1 | |||
AK296540.1 | ||||
AK303751.1 | ||||
AK314390.1 | ||||
AL096881.1 | CAB51405.1 | |||
BC058915.1 | AAH58915.1 | |||
CR456571.1 | CAG30457.1 | |||
DC375707.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss | MGC20647 RNF215 SF3A1 SLC35E4 TBC1D10A KIAA1656 SDC4P HORMAD2 SEC14L3 PES1 SEC14L4 GAL3ST1 OSM SEC14L6 TCN2 SEC14L2 GATSL3 CCDC157 MTFP1 LIF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |