Assay Details
Target Gene Details
Entrez Gene ID: | 23786 |
Gene Name: | BCL2 like 13 |
Gene Aliases: |
BCL-RAMBO, Bcl2-L-13, MIL1 |
Location: |
Chr.22:17628855-17730855 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv1008890 | Chr.22:17657148 - 17667590 on Build GRCh38 | Deletion | BCL2L13 |
nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss | SLC25A18 BCL2L13 CECR2 ATP6V1E1 LOC101929372 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | SLC25A18 BCL2L13 BID CECR2 MIR3198-1 ATP6V1E1 LINC00528 LOC101929372 MICAL3 |
esv3893429 | Chr.22:17600825 - 17678777 on Build GRCh38 | Gain | BCL2L13 ATP6V1E1 |
esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss | SLC25A18 MIR648 BCL2L13 BID MIR3198-1 ATP6V1E1 LINC00528 FLJ41941 LOC101929372 MICAL3 |
dgv4465n100 | Chr.22:17577691 - 17706530 on Build GRCh38 | Gain | SLC25A18 BCL2L13 ATP6V1E1 LOC101929372 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | TMEM191B MIR648 RIMBP3 MIR3198-1 LOC100996401 LOC642643 MICAL3 FAM230A SLC25A18 BCL2L13 BID LOC100996432 ATP6V1E1 LINC00528 LOC729461 PI4KAP1 USP18 CECR2 FLJ41941 LOC100996415 TUBA8 PEX26 GGT3P LOC102725072 LOC101929372 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |