Assay Details
Target Gene Details
Entrez Gene ID: | 4700 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A6 |
Gene Aliases: |
B14, CI-B14, LYRM6, NADHB14 |
Location: |
Chr.22:42085526-42090884 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA6 | NM_002490.3 | NP_002481.2 | ||
AF047182.1 | AAC04267.1 | |||
AK291874.1 | ||||
AK316562.1 | ||||
BC002772.1 | AAH02772.1 | |||
CR456529.1 | CAG30415.1 | |||
CR456714.1 | CAG32995.1 | |||
CR541997.1 | CAG46794.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508737 | Chr.22:42052007 - 42134336 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834210 | Chr.22:42057329 - 42209100 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)