Assay Details
Target Gene Details
Entrez Gene ID: | 164668 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 3H |
Gene Aliases: |
A3H, ARP-10, ARP10 |
Location: |
Chr.22:39097204-39104067 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOBEC3H | NM_001166002.2 | NP_001159474.2 | ||
NM_001166003.2 | NP_001159475.2 | |||
NM_001166004.2 | NP_001159476.2 | |||
NM_181773.4 | NP_861438.3 | |||
XM_011529990.2 | XP_011528292.1 | |||
XM_011529991.2 | XP_011528293.1 | |||
XM_011529992.2 | XP_011528294.1 | |||
BC069023.1 | AAH69023.1 | |||
BQ052182.1 | ||||
BQ054330.1 | ||||
BU585251.1 | ||||
CR456481.3 | CAG30367.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834197 | Chr.22:39077294 - 39214999 on Build GRCh38 | Gain | CBX7 APOBEC3H APOBEC3G |
nsv510807 | Chr.22:39075404 - 39147379 on Build GRCh38 | Deletion | CBX7 APOBEC3H APOBEC3G |
nsv834196 | Chr.22:38996564 - 39214999 on Build GRCh38 | Loss | CBX7 APOBEC3B-AS1 APOBEC3D APOBEC3H APOBEC3G APOBEC3F APOBEC3C |
nsv1066354 | Chr.22:39070438 - 39130755 on Build GRCh38 | Gain | APOBEC3H APOBEC3G |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |