Assay Details
Target Gene Details
Entrez Gene ID: | 3455 |
Gene Name: | interferon alpha and beta receptor subunit 2 |
Gene Aliases: |
IFN-R, IFN-alpha-REC, IFNABR, IFNARB, IMD45 |
Location: |
Chr.21:33229895-33264513 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IFNAR2 | NM_000874.4 | NP_000865.2 | ||
NM_001289125.1 | NP_001276054.1 | |||
NM_001289126.1 | NP_001276055.1 | |||
NM_001289128.1 | NP_001276057.1 | |||
NM_207584.2 | NP_997467.1 | |||
NM_207585.2 | NP_997468.1 | |||
BC002793.2 | AAH02793.1 | |||
BG715143.1 | ||||
CD110317.1 | ||||
HG380509.1 | ||||
HY104613.1 | ||||
L41942.1 | AAB46413.1 | |||
L41944.1 | AAB46415.1 | |||
U29584.1 | AAC50202.1 | |||
X77722.1 | CAA54785.1 | |||
X89814.1 | CAA61940.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |