Assay Details
Target Gene Details
Entrez Gene ID: | 539 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit |
Gene Aliases: |
ATPO, HMC08D05, OSCP |
Location: |
Chr.21:33903453-33915854 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP5O | NM_001697.2 | NP_001688.1 | ||
AF088071.1 | ||||
AK124187.1 | ||||
AK222608.1 | BAD96328.1 | |||
AK222962.1 | BAD96682.1 | |||
AK311796.1 | ||||
AV734214.1 | ||||
BC021233.2 | AAH21233.1 | |||
BC022865.1 | AAH22865.1 | |||
BT019836.1 | AAV38639.1 | |||
CD174837.1 | ||||
CR456822.1 | CAG33103.1 | |||
X83218.1 | CAA58219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv587399 | Chr.21:33834576 - 33969278 on Build GRCh38 | Gain | LINC00649 ITSN1 ATP5O |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |