Assay Details
Target Gene Details
Entrez Gene ID: | 56911 |
Gene Name: | MAP3K7 C-terminal like |
Gene Aliases: |
C21orf7, HC21ORF7, TAK1L, TAKL, TAKL-1, TAKL-2, TAKL-4 |
Location: |
Chr.21:29077471-29175889 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAP3K7CL | NM_001286617.1 | NP_001273546.1 | ||
NM_001286618.1 | NP_001273547.1 | |||
NM_001286619.1 | NP_001273548.1 | |||
NM_001286622.1 | NP_001273551.1 | |||
NM_001286623.1 | NP_001273552.1 | |||
NM_001286634.1 | NP_001273563.1 | |||
NM_020152.3 | NP_064537.1 | |||
AF269161.1 | AAF81751.1 | |||
AF269162.1 | AAF81752.1 | |||
AF269163.1 | AAF81753.1 | |||
AK122780.1 | ||||
AK294287.1 | ||||
AY171599.2 | AAO16519.1 | |||
BG566433.1 | ||||
EF219157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3646794 | Chr.21:29100737 - 29111625 on Build GRCh38 | Loss | MAP3K7CL |
nsv3485 | Chr.21:29068122 - 29112865 on Build GRCh38 | Deletion | CCT8 MAP3K7CL |
nsv3486 | Chr.21:29084659 - 29119954 on Build GRCh38 | Insertion | MAP3K7CL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |