Assay Details
Target Gene Details
Entrez Gene ID: | 571 |
Gene Name: | BTB domain and CNC homolog 1 |
Gene Aliases: |
BACH-1, BTBD24 |
Location: |
Chr.21:29298795-29361896 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BACH1 | NM_001186.3 | NP_001177.1 | ||
NM_206866.2 | NP_996749.1 | |||
AA402241.1 | ||||
AB002803.1 | BAA24932.1 | |||
AF026199.1 | AAB84100.1 | |||
AK291021.1 | ||||
AK313791.1 | ||||
BC063307.1 | AAH63307.1 | |||
DA855751.1 | ||||
HY028322.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |