Assay Details
Target Gene Details
Entrez Gene ID: | 2078 |
Gene Name: | ERG, ETS transcription factor |
Gene Aliases: |
erg-3, p55 |
Location: |
Chr.21:38367261-38662045 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERG | NM_001243432.2 | NP_001230361.1 | ||
AY204741.1 | AAP41719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834098 | Chr.21:38319976 - 38473331 on Build GRCh38 | Gain | LINC01423 ERG |
nsv1058886 | Chr.21:37969257 - 38427716 on Build GRCh38 | Gain | DSCR8 LOC100288590 LINC01423 KCNJ15 DSCR10 DSCR4 ERG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |