Assay Details
Target Gene Details
Entrez Gene ID: | 3588 |
Gene Name: | interleukin 10 receptor subunit beta |
Gene Aliases: |
CDW210B, CRF2-4, CRFB4, D21S58, D21S66, IL-10R2 |
Location: |
Chr.21:33266360-33297234 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL10RB | NM_000628.4 | NP_000619.3 | ||
AK124057.1 | BAC85761.1 | |||
AK299961.1 | ||||
AK313348.1 | ||||
AU133530.2 | ||||
BC001903.1 | AAH01903.1 | |||
BT009777.1 | AAP88779.1 | |||
JN591378.1 | ||||
Z17227.1 | CAA78933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv587376 | Chr.21:33271777 - 33307426 on Build GRCh38 | Gain | IL10RB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |