Assay Details
Target Gene Details
Entrez Gene ID: | 54148 |
Gene Name: | mitochondrial ribosomal protein L39 |
Gene Aliases: |
C21orf92, L39mt, MRP-L5, MRPL5, MSTP003, PRED22, PRED66, RPML5 |
Location: |
Chr.21:25585656-25607858 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPL39 | NM_017446.3 | NP_059142.2 | ||
NM_080794.3 | NP_542984.2 | |||
XM_006724026.3 | XP_006724089.1 | |||
XM_011529651.2 | XP_011527953.1 | |||
AF109357.1 | AAQ13505.1 | |||
AF239727.1 | AAF44696.1 | |||
AK000458.1 | BAA91177.1 | |||
AK225878.1 | ||||
BC004896.2 | AAH04896.2 | |||
BC107719.1 | AAI07720.1 | |||
BQ719968.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |