Assay Details
Target Gene Details
Entrez Gene ID: | 8624 |
Gene Name: | proteasome assembly chaperone 1 |
Gene Aliases: |
C21LRP, DSCR2, LRPC21, PAC-1, PAC1 |
Location: |
Chr.21:39175446-39183514 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PSMG1 | NM_001261824.1 | NP_001248753.1 | ||
NM_001320795.1 | NP_001307724.1 | |||
NM_003720.3 | NP_003711.1 | |||
NM_203433.2 | NP_982257.1 | |||
NR_049728.1 | ||||
XM_017028488.1 | XP_016883977.1 | |||
AA731200.1 | ||||
AB451468.1 | ||||
AF417108.1 | ||||
AJ006291.1 | CAA06957.1 | |||
AK001455.1 | ||||
AK308614.1 | ||||
AK315406.1 | ||||
AW594615.1 | ||||
AY463963.1 | AAR25628.1 | |||
BC003619.2 | AAH03619.1 | |||
BC010424.1 | AAH10424.1 | |||
BC011755.2 | AAH11755.1 | |||
BC012809.2 | AAH12809.1 | |||
BM013494.1 | ||||
BM462074.1 | ||||
BQ008489.1 | ||||
CR541821.1 | CAG46620.1 | |||
CR541852.1 | CAG46650.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)