Assay Details
Target Gene Details
Entrez Gene ID: | 58494 |
Gene Name: | junctional adhesion molecule 2 |
Gene Aliases: |
C21orf43, CD322, JAM-B, JAMB, PRO245, VE-JAM, VEJAM |
Location: |
Chr.21:25639282-25717562 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JAM2 | NM_001270407.1 | NP_001257336.1 | ||
NM_001270408.1 | NP_001257337.1 | |||
NM_021219.3 | NP_067042.1 | |||
NR_072999.1 | ||||
AF255910.1 | AAF81223.1 | |||
AK294769.1 | ||||
AK312708.1 | ||||
AX772824.1 | CAE06261.1 | |||
AY016009.1 | AAG49022.1 | |||
AY077698.1 | AAL82538.1 | |||
AY358361.1 | AAQ88727.1 | |||
BC017779.1 | AAH17779.1 | |||
BM804795.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)