Assay Details
Target Gene Details
Entrez Gene ID: | 100874042 |
Gene Name: | HM13 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:31567587-31573263 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HM13-AS1 | NR_046853.2 | |||
AJ710081.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470546 | Chr.20:31245806 - 31647499 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2763228 | Chr.20:31247246 - 31651719 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3645652 | Chr.20:31500710 - 31576564 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv3345 | Chr.20:31549053 - 31593946 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv458961 | Chr.20:31305698 - 31688103 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)