Assay Details
Target Gene Details
Entrez Gene ID: | 5770 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 1 |
Gene Aliases: |
PTP1B |
Location: |
Chr.20:50510321-50584762 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTPN1 | NM_001278618.1 | NP_001265547.1 | ||
NM_002827.3 | NP_002818.1 | |||
AK290638.1 | ||||
AK299830.1 | ||||
AK316563.1 | ||||
AU117677.1 | ||||
BC015660.2 | AAH15660.1 | |||
BC018164.1 | AAH18164.1 | |||
BT006752.1 | AAP35398.1 | |||
DC300354.1 | ||||
M31724.1 | AAA60223.1 | |||
M33689.1 | AAA60157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4324n100 | Chr.20:50512568 - 50535619 on Build GRCh38 | Gain | PTPN1 |
dgv163n111 | Chr.20:50509110 - 50534089 on Build GRCh38 | Duplication | PTPN1 |
nsv586177 | Chr.20:50358658 - 50546698 on Build GRCh38 | Gain | PTPN1 |
esv2760682 | Chr.20:50493794 - 50535631 on Build GRCh38 | Gain | PTPN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |