Assay Details
Target Gene Details
Entrez Gene ID: | 443 |
Gene Name: | aspartoacylase |
Gene Aliases: |
ACY2, ASP |
Location: |
Chr.17:3474106-3502534 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASPA | NM_000049.2 | NP_000040.1 | ||
NM_001128085.1 | NP_001121557.1 | |||
XM_017024661.1 | XP_016880150.1 | |||
AK312901.1 | ||||
BC029128.1 | AAH29128.1 | |||
CR541908.1 | CAG46706.1 | |||
S67156.1 | AAB29190.1 |
Target Gene Details
Entrez Gene ID: | 84690 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
NYD-SP20, NYDSP20 |
Location: |
Chr.17:3440011-3513852 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATA22 | NM_001321336.1 | NP_001308265.1 | ||
NM_001321337.1 | NP_001308266.1 | |||
AK295299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892950 | Chr.17:3131812 - 3577153 on Build GRCh38 | Loss | OR1E2 TRPV3 SPATA22 OR3A2 TRPV1 OR3A1 OR1D4 OR3A4P OR1A2 ASPA OR3A3 OR1E1 OR1A1 |
nsv574237 | Chr.17:3486822 - 3729542 on Build GRCh38 | Loss | GSG2 EMC6 TRPV3 SPATA22 TAX1BP3 ASPA ITGAE CTNS P2RX5-TAX1BP3 P2RX5 SHPK TRPV1 |
nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain | PAFAH1B1 EMC6 TRPV3 TAX1BP3 OR1D2 CLUH TRPV1 MIR1253 CCDC92B OR3A1 MIR6776 OR1D5 CTNS P2RX1 ATP2A3 NCBP3 P2RX5-TAX1BP3 SHPK OR1E1 OR1A1 LOC105371592 RAP1GAP2 CAMKK1 OR1E2 SPATA22 LOC101927911 P2RX5 OR3A2 GSG2 LOC105371591 OR1D4 OR3A4P OR1A2 ASPA ITGAE OR1G1 OR3A3 |
nsv574232 | Chr.17:3254715 - 3503170 on Build GRCh38 | Gain | OR3A1 OR1E2 OR3A4P SPATA22 ASPA OR3A3 OR3A2 OR1E1 |
esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion | PAFAH1B1 LOC105371592 RAP1GAP2 OR1E2 TRPV3 SPATA22 LOC101927911 OR1D2 CLUH OR3A2 MIR1253 CCDC92B OR3A1 LOC105371591 OR1D4 OR3A4P MIR6776 OR1A2 OR1D5 ASPA OR1G1 OR3A3 OR1E1 OR1A1 |
nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain | CAMKK1 OR1E2 EMC6 TRPV3 SPATA22 TAX1BP3 P2RX5 OR3A2 TRPV1 GSG2 ASPA ITGAE CTNS OR3A3 NCBP3 P2RX5-TAX1BP3 SHPK OR1E1 |
nsv528368 | Chr.17:3459775 - 3507608 on Build GRCh38 | Gain | SPATA22 ASPA |
nsv833341 | Chr.17:3352243 - 3521390 on Build GRCh38 | Loss | OR1E2 TRPV3 SPATA22 ASPA OR3A3 OR3A2 OR1E1 |
nsv1060071 | Chr.17:3398474 - 3633472 on Build GRCh38 | Loss | OR1E2 TRPV3 SPATA22 ASPA OR3A3 SHPK TRPV1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |