Assay Details
Target Gene Details
Entrez Gene ID: | 23729 |
Gene Name: | sedoheptulokinase |
Gene Aliases: |
CARKL, SHK |
Location: |
Chr.17:3608262-3636322 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHPK | NM_013276.2 | NP_037408.2 | ||
AF163573.1 | AAF24936.1 | |||
AK055609.1 | ||||
AK298360.1 | ||||
AK312428.1 | ||||
AL832420.1 | CAH10646.1 | |||
BC020543.1 | AAH20543.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3085n100 | Chr.17:3595117 - 3665113 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3639749 | Chr.17:3570544 - 3639124 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3639752 | Chr.17:3601104 - 3658235 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv882e212 | Chr.17:3585851 - 3654158 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3572354 | Chr.17:3528675 - 3654158 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv2762588 | Chr.17:3602274 - 3654159 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv5423n54 | Chr.17:3601261 - 3665107 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv516676 | Chr.17:3553441 - 3652130 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
nsv574237 | Chr.17:3486822 - 3729542 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2662762 | Chr.17:3601001 - 3658179 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv526175 | Chr.17:3528392 - 3616421 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv358n27 | Chr.17:3601921 - 3664167 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv1060071 | Chr.17:3398474 - 3633472 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs76056301] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)