Assay Details
Target Gene Details
Entrez Gene ID: | 10948 |
Gene Name: | StAR related lipid transfer domain containing 3 |
Gene Aliases: |
CAB1, MLN64, es64 |
Location: |
Chr.17:39637080-39664201 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD3 | NM_001165937.1 | NP_001159409.1 | ||
NM_001165938.1 | NP_001159410.1 | |||
NM_006804.3 | NP_006795.3 | |||
XM_006721646.2 | XP_006721709.1 | |||
XM_011524210.1 | XP_011522512.1 | |||
XM_011524213.1 | XP_011522515.1 | |||
AK225069.1 | ||||
AK300176.1 | ||||
AK300842.1 | ||||
AK301524.1 | ||||
AK303114.1 | ||||
BC008356.1 | AAH08356.1 | |||
BC008747.2 | AAH08747.1 | |||
BC025679.1 | AAH25679.1 | |||
D38255.1 | BAA22525.1 | |||
DC367486.1 | ||||
X80198.1 | CAA56489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521294 | Chr.17:39538456 - 39648605 on Build GRCh38 | Loss | PPP1R1B NEUROD2 CDK12 STARD3 |
nsv516757 | Chr.17:39625596 - 39678288 on Build GRCh38 | Loss | PGAP3 PNMT PPP1R1B TCAP STARD3 |
nsv574991 | Chr.17:39613752 - 39674782 on Build GRCh38 | Loss | PGAP3 PNMT PPP1R1B TCAP STARD3 |
dgv40n68 | Chr.17:39536627 - 39726611 on Build GRCh38 | Loss | MIR4728 PGAP3 PNMT PPP1R1B TCAP NEUROD2 CDK12 ERBB2 STARD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |