Assay Details
Target Gene Details
Entrez Gene ID: | 8711 |
Gene Name: | tyrosine kinase non receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.17:7380559-7389774 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNK1 | NM_001251902.1 | NP_001238831.1 | ||
NM_003985.4 | NP_003976.2 | |||
XM_011524045.2 | XP_011522347.1 | |||
XM_011524046.2 | XP_011522348.1 | |||
XM_011524047.2 | XP_011522349.1 | |||
XM_011524048.1 | XP_011522350.1 | |||
AK225403.1 | ||||
AK310684.1 | ||||
BC035782.1 | AAH35782.1 | |||
U43408.1 | AAC50427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion | DVL2 SHBG ZBTB4 SLC16A11 PHF23 EIF5A MIR497 EIF4A1 MIR497HG TEKT1 LOC100996842 SAT2 TMEM102 KCTD11 NEURL4 DLG4 SLC2A4 SLC16A13 TNK1 CHRNB1 SOX15 TMEM95 CLEC10A ASGR2 SENP3-EIF4A1 ALOX12P2 YBX2 RNASEK ASGR1 SNORA48 FGF11 FXR2 SENP3 CD68 CTDNEP1 SNORD10 SNORA67 C17orf74 ELP5 TMEM256 ALOX12 CLDN7 MPDU1 TNFSF12 PLSCR3 SLC35G6 GPS2 NLGN2 BCL6B MIR195 POLR2A ACADVL ACAP1 SPEM1 FBXO39 GABARAP TNFSF12-TNFSF13 MIR324 ALOX12-AS1 ATP1B2 TNFSF13 C17orf49 RNASEK-C17orf49 LOC107983988 TMEM256-PLSCR3 |
nsv523672 | Chr.17:7379825 - 7549660 on Build GRCh38 | Loss | SLC35G6 NLGN2 TMEM102 FGF11 POLR2A SPEM1 TNFSF12-TNFSF13 TNK1 CHRNB1 ZBTB4 C17orf74 TMEM256 TNFSF12 PLSCR3 LOC107983988 TMEM256-PLSCR3 |
nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion | FGF11 DVL2 CTDNEP1 C17orf74 ELP5 PHF23 TMEM256 EIF5A CLDN7 PLSCR3 GPS2 NLGN2 TMEM102 KCTD11 NEURL4 DLG4 ACADVL SLC2A4 ACAP1 SPEM1 GABARAP TNK1 CHRNB1 MIR324 TMEM95 YBX2 ASGR1 LOC107983988 TMEM256-PLSCR3 |
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | DVL2 SHBG ZBTB4 SLC16A11 PHF23 EIF5A MIR497 EIF4A1 MIR497HG LOC100996842 SAT2 TMEM102 KCTD11 NEURL4 DLG4 SLC2A4 SLC16A13 TNK1 CHRNB1 SOX15 TMEM95 CLEC10A ASGR2 SENP3-EIF4A1 YBX2 RNASEK ASGR1 SNORA48 FGF11 FXR2 SENP3 CD68 CTDNEP1 SNORD10 SNORA67 C17orf74 ELP5 TMEM256 ALOX12 CLDN7 MPDU1 TNFSF12 PLSCR3 SLC35G6 GPS2 NLGN2 BCL6B MIR195 POLR2A ACADVL ACAP1 SPEM1 GABARAP TNFSF12-TNFSF13 MIR324 WRAP53 TP53 ALOX12-AS1 ATP1B2 TNFSF13 C17orf49 RNASEK-C17orf49 LOC107983988 TMEM256-PLSCR3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |