Assay Details
Target Gene Details
Entrez Gene ID: | 100130950 |
Gene Name: | uncharacterized LOC100130950 |
Gene Aliases: |
- |
Location: |
Chr.17:5192084-5235636 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100130950 | NR_034082.1 |
Target Gene Details
Entrez Gene ID: | 388325 |
Gene Name: | SLP adaptor and CSK interacting membrane protein |
Gene Aliases: |
C17orf87, UNQ5783 |
Location: |
Chr.17:5208920-5234860 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCIMP | NM_001271842.1 | NP_001258771.1 | ||
NM_001319190.1 | NP_001306119.1 | |||
NM_207103.3 | NP_996986.1 | |||
XM_017024615.1 | XP_016880104.1 | |||
AK057142.1 | ||||
AK297035.1 | ||||
AK310145.1 | ||||
AY358809.1 | AAQ89169.1 | |||
BC127925.1 | ||||
BC130554.1 | ||||
BC130556.1 | ||||
BC144395.1 | ||||
DQ778084.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain | ENO3 LOC107984973 RABEP1 SCIMP LOC101559451 C17orf107 C1QBP GLTPD2 ARRB2 LOC101927979 VMO1 ZFP3 RPAIN SMTNL2 LOC100130950 CXCL16 MINK1 DHX33 MIR6865 LOC102724009 MED11 ZNF232 CHRNE LOC101928000 SPAG7 RNF167 ALOX15 NUP88 PFN1 INCA1 PELP1 SLC52A1 GP1BA USP6 KIF1C TM4SF5 ZMYND15 LOC400568 PSMB6 ZNF594 CAMTA2 PLD2 MIR6864 SLC25A11 |
dgv3089n100 | Chr.17:5187013 - 5327634 on Build GRCh38 | Gain | RABEP1 ZNF594 SCIMP LOC100130950 |
nsv952114 | Chr.17:5228306 - 5237005 on Build GRCh38 | Deletion | SCIMP LOC100130950 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |